missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylmethylamine, 98%, Thermo Scientific Chemicals

Product Code. 10294470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10294470 100 mL Glass bottle 100mL
10386030 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10294470 Supplier Thermo Scientific Acros Supplier No. 114461000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-39-7
Molecular Formula C5H14N
Molecular Weight (g/mol) 88.17
MDL Number MFCD00009049
InChI Key GNVRJGIVDSQCOP-UHFFFAOYSA-O
Synonym n,n-diethylmethylamine, diethylmethylamine, ethanamine, n-ethyl-n-methyl, n-methyldiethylamine, diethylamine, n-methyl, n-ethyl-n-methyl-ethanamine, methyldiethylamine, diethylmethyl amine, diethyl methyl amine, methyl diethyl amine
PubChem CID 12022
IUPAC Name N-ethyl-N-methylethanamine
SMILES CC[NH+](C)CC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -196°C
Color Colorless to Yellow
Density 0.7200g/mL
Boiling Point 63°C to 65°C
Flash Point -23°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C2H5)2NCH3
Refractive Index 1.3890 to 1.391
Quantity 100 mL
Beilstein 04, 99
Specific Gravity 0.72
Solubility Information Solubility in water: soluble. Other solubilities: very soluble in ethanol and ethyl ether,soluble in most common organic solvents
Formula Weight 87.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N, N-Diethylmethylamine, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylmethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.