missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylhydrazine dihydrochloride, tech. 90%

Product Code. 11456006
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456006 5 g 5g
11466006 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456006 Supplier Thermo Scientific Alfa Aesar Supplier No. B22317.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7699-31-2 | C4H14Cl2N2 | 161.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7699-31-2
Molecular Formula C4H14Cl2N2
Molecular Weight (g/mol) 161.07
MDL Number MFCD00012523
InChI Key NMTDWUVFGOBBAX-UHFFFAOYSA-N
Synonym 1,2-diethylhydrazine dihydrochloride, n,n'-diethylhydrazine dihydrochloride, unii-x3o3iwe4tg, ccris 8919, hydrazine, 1,2-diethyl-, dihydrochloride, x3o3iwe4tg, 1,2-diethylhydrazine 2hcl, diethylhydrazine dihydrochloride, 1,2-diethyl hydrazine dihydrochloride, 1,2-diethylhydrazine dihydrochlo-ride
PubChem CID 24372
IUPAC Name 1,2-diethylhydrazine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].CCNNCC

Specifications

Melting Point ∼169°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3668380
Formula Weight 159.06
Percent Purity 90%
Grade Technical
Chemical Name or Material N,N'-Diethylhydrazine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.