missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethylenediamine, 99%

Product Code. 11414187
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414187 500 g 500g
11404187 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11414187 Supplier Thermo Scientific Alfa Aesar Supplier No. A19636.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-36-7 | C6H16N2 | 116.208 g/mol

N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides. It is also used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylethylenediamine was used in the oxidative amidation of hydrogen phosphonate diesters in the synthesis of oligonucleotides. It is also used to develop melanin targeted PET and single-photon emission computed tomography (SPECT) imaging agents for melanoma.

Solubility
Miscible in water.

Notes
Air sensitive. Store away from oxidizing agents. Protect against electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-36-7
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008176
InChI Key UDGSVBYJWHOHNN-UHFFFAOYSA-N
Synonym n,n-diethylethylenediamine, 2-diethylamino ethylamine, n,n-diethyl-1,2-ethanediamine, diethylamino ethylamino, n1,n1-diethylethane-1,2-diamine, 2-aminoethyl diethylamine, 2-aminoethyldiethylamine, n,n-diethylethanediamine, ethylenediamine, n,n-diethyl, n,n-diethylethane-1,2-diamine
PubChem CID 60993
IUPAC Name N',N'-diethylethane-1,2-diamine
SMILES CCN(CC)CCN

Specifications

Melting Point <-70°C
Density 0.825
Boiling Point 145°C to 147°C
Flash Point 30°C (86°F)
Refractive Index 1.436
Quantity 500 g
UN Number UN2685
Beilstein 605299
Solubility Information Miscible in water.
Formula Weight 116.21
Percent Purity 99%
Chemical Name or Material N,N-Diethylethylenediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Diethylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Diethylethylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.