missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylethylenediamine, 96%

Product Code. 11498986
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498986 5 g 5g
11408996 25 g 25g
11418996 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498986 Supplier Thermo Scientific Alfa Aesar Supplier No. A17113.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-74-0 | C6H16N2 | 116.21 g/mol

N,N'-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N'-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used as an internal standard in liquid chromatographic method used for checking the assay of ethambutol. It is utilized for the solvation of lithium hexamethyldisilazide. It acts as a ligand and form coordination complexes such as trans-diaquabis(N,N′-diethylethylenediamine)nickel(II) chloride and copper(II)-N,N-Diethylethylenediamine, which shows thermochromic properties. Further, it is used as a pharmaceutical intermediate used in the synthesis of procainamide hydrochloride, tiapride and cinchocaine.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-74-0
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00009033
InChI Key CJKRXEBLWJVYJD-UHFFFAOYSA-N
Synonym n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide
PubChem CID 67105
ChEBI CHEBI:182290
IUPAC Name N,N'-diethylethane-1,2-diamine
SMILES CCNCCNCC

Specifications

Density 0.811
Boiling Point 152°C to 154°C
Flash Point 33°C (92°F)
Refractive Index 1.4326
Quantity 5 g
UN Number UN2685
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 116.21
Percent Purity 96%
Chemical Name or Material N,N'-Diethylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.