missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylethylenediamine, 95%

Product Code. 15274644 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15274644 5 g Glass bottle 5g
15284644 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15274644 Supplier Thermo Scientific Acros Supplier No. 155520050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-74-0 | C6H16N2 | 116.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-74-0
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.21
MDL Number MFCD00009033
InChI Key CJKRXEBLWJVYJD-UHFFFAOYSA-N
Synonym n,n'-diethylethylenediamine, 1,2-ethanediamine, n,n'-diethyl, 1,2-bis ethylamino ethane, 3,6-diazaoctane, 1,2-ethanediamine, n1,n2-diethyl, n,n'-ethylenediethyldiamine, ethyl 2-ethylamino ethyl amine, ethylenediamine, n,n'-diethyl, n,n'-diethyl-1,2-diaminoethane, dihydrobromide
PubChem CID 67105
ChEBI CHEBI:182290
IUPAC Name N,N'-diethylethane-1,2-diamine
SMILES CCNCCNCC

Specifications

Density 0.8110g/mL
Boiling Point 152.0°C to 154.0°C
Flash Point 33°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C2H5NHCH2CH2NHC2H5
Refractive Index 1.4326
Quantity 5 g
Beilstein 04,251
Specific Gravity 0.811
Solubility Information Solubility in water: miscible.
Formula Weight 116.21
Percent Purity 95%
Chemical Name or Material N, N'-Diethylethylenediamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N'-Diethylethylenediamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.