missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diethylethanol-1,1,2,2-d4-amine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-(Diethylamino)ethanol-1,1,2,2-D4 |
| CAS | 2733149-33-0 |
| Molecular Formula | C6 D4 H11 N O |
| Quantity | 0.25 g |
| Synonym | N,N-Diethylethanol-1,1,2,2-d4-amine, 2-(Diethylamino)ethanol-1,1,2,2-d4, 2-(Diethylamino)ethanol-1,1,2,2-D4 |
| InChI Formula | InChI=1 S/C6H15NO/c1-3-7(4-2)5-6-8/h8H,3-6H2,1-2H3/i5D2,6D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])N(CC)CC |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(diethylamino)ethanol |
| Molecular Weight (g/mol) | 121.214 |
| Formula Weight | 121.14 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction