missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylaniline, 99%, Thermo Scientific Chemicals

Product Code. 10477940
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-66-7
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00009042,MFCD31699978
InChI Key GGSUCNLOZRCGPQ-UHFFFAOYSA-N
Synonym diethylaniline, n,n-diethylanilin, diethylphenylamine, benzenamine, n,n-diethyl, n,n-diethyl aniline, diaethylanilin, phenyldiethylamine, aniline, n,n-diethyl, n,n-diethylaminobenzene, n-phenyldiethylamine
PubChem CID 7061
IUPAC Name N,N-diethylaniline
SMILES CCN(CC)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -38.0°C
Color Yellow
Density 0.9330g/mL
Boiling Point 215.0°C to 217.0°C
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H5N(C2H5)2
Refractive Index 1.5410 to 1.5430
Quantity 250 mL
Beilstein 12, 164
Fieser 05,212; 16,182
Merck Index 15, 3125
Specific Gravity 0.933
Solubility Information Solubility in water: 10-27g/L. Other solubilities: slightly soluble in alcohol,chloroform and ether
Formula Weight 149.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Diethylaniline, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'