missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylacetoacetamide, 97%

Product Code. 11446936
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446936 50 g 50g
11456936 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446936 Supplier Thermo Scientific Alfa Aesar Supplier No. B24019.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2235-46-3 | C8H15NO2 | 157.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2235-46-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00026728
InChI Key NTMXFHGYWJIAAE-UHFFFAOYSA-N
Synonym n,n-diethylacetoacetamide, n,n-diethyl-3-oxobutyramide, diethylacetoacetamide, butanamide, n,n-diethyl-3-oxo, acetoacetamide, n,n-diethyl, 1-diethylcarbamoyl-2-propanone, n,n-diethylacetylacetamide, acetoacetdiethylamide, diethylamid kyseliny acetoctove, diethylamid kyseliny acetoctove czech
PubChem CID 16699
IUPAC Name N,N-diethyl-3-oxobutanamide
SMILES CCN(CC)C(=O)CC(=O)C

Specifications

Melting Point <-73°C
Density 0.99
Boiling Point 213°C to 218°C
Flash Point 94°C (201°F)
Refractive Index 1.472
Quantity 50 g
Beilstein 636486
Formula Weight 157.2
Percent Purity 97%
Chemical Name or Material N,N-Diethylacetoacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.