missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl(trimethylsilyl)amine, TRC

Product Code. 30382568
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
30382568 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 30382568

Brand: Toronto Research Chemicals TRCD4452655G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Diethyl(trimethylsilyl)amine
CAS 996-50-9
Molecular Formula C7H19NSi
Quantity 5 g
Synonym N,N-Diethyl-1,1,1-trimethyl-silylamine, (Diethylamino)trimethylsilane, (N,N-Diethylamino)trimethylsilane, (Trimethylsilyl)diethylamine, Diethyl(trimethylsilyl)amine, N-Ethyl-N-(trimethylsilyl)ethanamine,, N,N-Diethyl-1,1,1-trimethylsilanamine, N,N-Diethyl-1,1,1-trimethylsilylamine, N,N-Diethyltrimethylsilanamine, N,N-Diethyltrimethylsilylamine, N-(Trimethylsilyl)diethylamine, NSC 377650, Trimethyl(diethylamino)silane
InChI Formula InChI=1S/C7H19NSi/c1-6-8(7-2)9(3,4)5/h6-7H2,1-5H3
SMILES CCN(CC)[Si](C)(C)C
IUPAC Name N-ethyl-N-trimethylsilylethanamine
Molecular Weight (g/mol) 145.32
Formula Weight 145.1287
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.