missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl(trimethylsilyl)amine, TRC

Product Code. 30374899
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30374899 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30374899 Supplier Toronto Research Chemicals Supplier No. TRCD445265100G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material N,N-Diethyl(trimethylsilyl)amine
CAS 996-50-9
Molecular Formula C7H19NSi
Quantity 100 g
Synonym N,N-Diethyl-1,1,1-trimethyl-silylamine, (Diethylamino)trimethylsilane, (N,N-Diethylamino)trimethylsilane, (Trimethylsilyl)diethylamine, Diethyl(trimethylsilyl)amine, N-Ethyl-N-(trimethylsilyl)ethanamine,, N,N-Diethyl-1,1,1-trimethylsilanamine, N,N-Diethyl-1,1,1-trimethylsilylamine, N,N-Diethyltrimethylsilanamine, N,N-Diethyltrimethylsilylamine, N-(Trimethylsilyl)diethylamine, NSC 377650, Trimethyl(diethylamino)silane
InChI Formula InChI=1S/C7H19NSi/c1-6-8(7-2)9(3,4)5/h6-7H2,1-5H3
SMILES CCN(CC)[Si](C)(C)C
IUPAC Name N-ethyl-N-trimethylsilylethanamine
Molecular Weight (g/mol) 145.32
Formula Weight 145.1287
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.