missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-toluamide, TRC

Product Code. 30240670
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240670 50 mg 50mg
30240604 250 mg 250mg
30240534 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240670 Supplier Toronto Research Chemicals Supplier No. TRCD47811550MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2728-05-4
Molecular Formula C12 H17 N O
Molecular Weight (g/mol) 191.27
Synonym N,N-Diethyl-4-toluamide
IUPAC Name N,N-diethyl-4-methylbenzamide
SMILES CCN(CC)C(=O)c1ccc(C)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H17NO/c1-4-13(5-2)12(14)11-8-6-10(3)7-9-11/h6-9H,4-5H2,1-3H3
Formula Weight 191.131
Chemical Name or Material N,N-Diethyl-4-toluamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.