missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine sulfate, 99%

Product Code. 10085053 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10085053 1 kg Glass bottle 1kg
10197133 50 g Glass bottle 50g
10157183 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10085053 Supplier Thermo Scientific Acros Supplier No. 395000010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

N, N-Diethyl-p-phenylenediamine sulfate, 99%, C10H16N2.H2SO4, CAS Number-6283-63-2 | CAS: 6283-63-2 | C10H16N2·H2SO4 | 262.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6283-63-2
Molecular Formula C10H16N2·H2SO4
Molecular Weight (g/mol) 262.33
MDL Number MFCD00012993
InChI Key AYLDJQABCMPYEN-UHFFFAOYSA-N
Synonym n,n-diethyl-p-phenylenediamine sulfate, n1,n1-diethylbenzene-1,4-diamine sulfate, 4-amino-n,n-diethylaniline sulfate, diethyl-p-phenylenediamine sulfate, n,n-diethyl-1,4-phenylenediamine sulfate, unii-usp19t3gda, 1,4-benzenediamine, n,n-diethyl-, sulfate 1:1, 1,4-benzenediamine, n,n-diethyl-, sulfate, n,n-diethyl-1,4-benzenediamine sulfate, p-phenylenediamine, n,n-diethyl-, sulfate 1:1
PubChem CID 80166
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine;sulfuric acid
SMILES CCN(CC)C1=CC=C(C=C1)N.OS(=O)(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 184.0°C to 187.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 kg
Formula Weight 262.33
Percent Purity 99%
Physical Form Crystalline Powder or Crystals With Lumps
Chemical Name or Material N, N-Diethyl-p-phenylenediamine sulfate
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Diethyl-p-phenylenediamine sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.