missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine oxalate, 96%

Product Code. 10107040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10107040 25 g Glass bottle 25g
10769901 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10107040 Supplier Thermo Scientific Acros Supplier No. 407410250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62637-92-7 | 0·5 C2H2O4 | 209.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62637-92-7
Molecular Formula 0·5 C2H2O4
Molecular Weight (g/mol) 209.27
MDL Number MFCD00070242

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 145.0°C to 150.0°C
Color Brown to Gray or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 25 g
Formula Weight 209.27
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Diethyl-p-phenylenediamine oxalate, 96%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Diethyl-p-phenylenediamine oxalate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.