missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-p-phenylenediamine, 98%

Product Code. 10741325 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10741325 500 g Glass bottle 500g
10025710 5 g Glass bottle 5g
10206061 25 g Glass bottle 25g
10435675 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10741325 Supplier Thermo Scientific Acros Supplier No. 407395000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-05-0 | C10H16N2 | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-05-0
MDL Number MFCD00007861
InChI Key QNGVNLMMEQUVQK-UHFFFAOYSA-N
Synonym n,n-diethyl-1,4-phenylenediamine, 4-amino-n,n-diethylaniline, n,n-diethyl-p-phenylenediamine, n1,n1-diethylbenzene-1,4-diamine, p-aminodiethylaniline, 4-diethylamino aniline, p-amino-n,n-diethylaniline, n,n-diethyl-1,4-benzenediamine, diethylaminoaniline, p-diethylamino aniline
PubChem CID 7120
IUPAC Name 4-N,4-N-diethylbenzene-1,4-diamine
SMILES CCN(CC)C1=CC=C(C=C1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 23.0°C
Density 0.9880g/mL
Boiling Point 268.0°C
Flash Point 139°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H16N2
Linear Formula (CH3CH2)2NC6H4NH2
Refractive Index 1.5690 to 1.5730 (20°C, 589nm)
Quantity 500 g
Specific Gravity 0.988
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Molecular Weight (g/mol) 164.25
Formula Weight 164.25
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material N, N-Diethyl-p-phenylenediamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethyl-p-phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.