missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium Imidodisulfuryl Fluoride, TRC

Product Code. 30373466
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373466 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30373466 Supplier Toronto Research Chemicals Supplier No. TRCD67908010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Diethyl-N-methyl-N-(2-methoxyethyl)ammonium Imidodisulfuryl Fluoride
CAS 1079129-48-8
Molecular Formula C8 H20 N O . F2 N O4 S2
Quantity 10 mg
InChI Formula InChI=1S/C8H20NO.F2NO4S2/c1-5-9(3,6-2)7-8-10-4;1-8(4,5)3-9(2,6)7/h5-8H2,1-4H3;/q+1;-1
SMILES CC[N+](C)(CC)CCOC.FS(=O)(=O)[N-]S(=O)(=O)F
IUPAC Name bis(fluorosulfonyl)azanide;diethyl-(2-methoxyethyl)-methylazanium
Molecular Weight (g/mol) 326.382
Formula Weight 326.078
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.