missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-N’-1-naphthylethylenediamine Oxalate, TRC

Product Code. 30386246
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386246 1 g
30376991 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386246 Supplier Toronto Research Chemicals Supplier No. TRCD4437351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29473-53-8
Molecular Formula C18H24N2O4
Molecular Weight (g/mol) 332.39
Synonym N1,N1-Diethyl-N2-1-naphthalenyl-1,2-ethanediamine Ethanedioate, β-Diethylaminoethyl-α-naphthylamine Oxalate
SMILES CCN(CC)CCNC1=CC=CC2=CC=CC=C21.C(=O)(C(=O)O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H22N2.C2H2O4/c1-3-18(4-2)13-12-17-16-11-7-9-14-8-5-6-10-15(14)16;3-1(4)2(5)6/h5-11,17H,3-4,12-13H2,1-2H3;(H,3,4)(H,5,6)
Formula Weight 332.1736
Chemical Name or Material N,N-Diethyl-N'-1-naphthylethylenediamine Oxalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.