missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-3-pyrrolidinamine Dihydrochloride, TRC

Product Code. 30383166
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383166 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383166 Supplier Toronto Research Chemicals Supplier No. TRCD49624810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Diethyl-3-pyrrolidinamine Dihydrochloride
CAS 20984-82-1
Molecular Formula C8H18N2 • 2(HCl)
Quantity 10 mg
InChI Formula InChI=1S/C8H18N2.ClH/c1-3-10(4-2)8-5-6-9-7-8;/h8-9H,3-7H2,1-2H3;1H
SMILES Cl.N1CCC(N(CC)CC)C1
IUPAC Name N,N-diethylpyrrolidin-3-amine hydrochloride
Molecular Weight (g/mol) 215.16
Formula Weight 214.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.