missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-3-methylbenzamide, 97%

Product Code. p-7023683
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11418097

Brand: Thermo Scientific Alfa Aesar A17486.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 134-62-3 | C12H17NO | 191.274 g/mol

It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most common ingredient in insect repellants and is believed to work as such in that mosquitoes intensely dislike its smell. Deet is also an effective solvent and may dissolve plastics, rayon, spandex, other synthetic fabrics and painted or varnished surfaces.

Solubility
Soluble in water.

Notes
Keep away from strong oxidizing agents, strong acids, strong bases, strong reducing agents. Keep ate room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-62-3
Molecular Formula C12H17NO
Molecular Weight (g/mol) 191.274
MDL Number MFCD00009046
InChI Key MMOXZBCLCQITDF-UHFFFAOYSA-N
Synonym deet, n,n-diethyl-m-toluamide, diethyltoluamide, dieltamid, metadelphene, flypel, delphene, deta, detamide, autan
PubChem CID 4284
ChEBI CHEBI:7071
IUPAC Name N,N-diethyl-3-methylbenzamide
SMILES CCN(CC)C(=O)C1=CC(=CC=C1)C

Specifications

Density 0.997
Boiling Point 110°C to 112°C (1 mmHg)
Flash Point 145°C (293°F)
Refractive Index 1.523
Quantity 100 g
Beilstein 2046711
Merck Index 14,2856
Solubility Information Soluble in water.
Formula Weight 191.27
Percent Purity 97%
Chemical Name or Material N,N-Diethyl-3-methylbenzamide
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Diethyl-3-methylbenzamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Diethyl-m-toluamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.