missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2-hydroxyacetamide, TRC

Product Code. 30382144
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382144 250 mg
30384018 1 g
30373443 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382144 Supplier Toronto Research Chemicals Supplier No. TRCD444580250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39096-01-0
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.17
Synonym 2-Hydroxy-N,N-diethylacetamide, N,N-Diethylglycolamide, N,N-diethylglycolamide
IUPAC Name N,N-diethyl-2-hydroxyacetamide
SMILES CCN(CC)C(=O)CO

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C6H13NO2/c1-3-7(4-2)6(9)5-8/h8H,3-5H2,1-2H3
Formula Weight 131.0946
Chemical Name or Material N,N-Diethyl-2-hydroxyacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.