missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2-fluoro-6-nitroaniline, 97%, Thermo Scientific™

£16.35 - £190.00

Chemical Identifiers

CAS 1873687-86-5
Synonym Diethyl-(2-fluoro-6-nitrophenyl)amine
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15489608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63104.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15469608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63104.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15479608
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63104.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1873687-86-5
Diethyl-(2-fluoro-6-nitrophenyl)amine

Specifications

1873687-86-5
Diethyl-(2-fluoro-6-nitrophenyl)amine
97%
N,N-Diethyl-2-fluoro-6-nitroaniline
UN2811
212.22
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications