missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2,2,2-trifluoroacetamide, TRC

Product Code. 30376636
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376636 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376636 Supplier Toronto Research Chemicals Supplier No. TRCD679295250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Diethyl-2,2,2-trifluoroacetamide
CAS 360-92-9
Molecular Formula C6H10F3NO
Quantity 250 mg
Synonym N,N-Diethyltrifluoroacetamide, NSC 14815, Trifluoroacetyl Diethylamine
InChI Formula InChI=1S/C6H10F3NO/c1-3-10(4-2)5(11)6(7,8)9/h3-4H2,1-2H3
SMILES CCN(CC)C(=O)C(F)(F)F
IUPAC Name N,N-diethyl-2,2,2-trifluoroacetamide
Molecular Weight (g/mol) 169.14
Formula Weight 169.0714
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.