missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine, 90%

Product Code. 10435095 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10435095 100 g Glass bottle 100g
10752284 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10435095 Supplier Thermo Scientific Acros Supplier No. 389211000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 309-88-6 | C7H11F6N | 223.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 309-88-6
Molecular Formula C7H11F6N
Molecular Weight (g/mol) 223.16
MDL Number MFCD00054683
InChI Key BNTFCVMJHBNJAR-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant
PubChem CID 136149
IUPAC Name N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine
SMILES CCN(CC)C(C(C(F)(F)F)F)(F)F

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2300g/mL
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 90% (Sum of two isomers) Approximate composition (GC)
Packaging Glass bottle
Linear Formula (CH3CH2)2NCH2F2CHFCF3
Quantity 100 g
Merck Index 15, 5153
Specific Gravity 1.23
Formula Weight 223.16
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.