Learn More
N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine, 90%, Thermo Scientific Chemicals
£52.30 - £163.00
Chemical Identifiers
CAS | 309-88-6 |
---|---|
Molecular Formula | C7H11F6N |
Molecular Weight (g/mol) | 223.16 |
MDL Number | MFCD00054683 |
InChI Key | BNTFCVMJHBNJAR-UHFFFAOYSA-N |
Synonym | n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant |
PubChem CID | 136149 |
IUPAC Name | N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine |
SMILES | CCN(CC)C(C(C(F)(F)F)F)(F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
309-88-6 | |
223.16 | |
BNTFCVMJHBNJAR-UHFFFAOYSA-N | |
136149 | |
CCN(CC)C(C(C(F)(F)F)F)(F)F |
C7H11F6N | |
MFCD00054683 | |
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant | |
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine |
Specifications
309-88-6 | |
100.0 | |
1.2300g/mL | |
Authentic | |
C7H11F6N | |
MFCD00054683 | |
1.23 | |
BNTFCVMJHBNJAR-UHFFFAOYSA-N | |
N,N-diethyl-1,1,2,3,3,3-hexafluoropropan-1-amine | |
136149 | |
90% | |
N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine |
88.0 | |
Colorless to Yellow | |
40°C | |
90% (Sum of two isomers) Approximate composition (GC) | |
(CH3CH2)2NCH2F2CHFCF3 | |
15, 5153 | |
n,n-diethyl-1,1,2,3,3,3-hexafluoropropylamine, n,n-diethyl 1,1,2,3,3,3-hexafluoropropylamine, diethyl 1,1,2,3,3,3-hexafluoropropyl amine, 1,1,2,3,3,3-hexafluoro-1-diethylaminopropane, n,n-diethyl-1,1,2,3,3,3-hexafluoro-1-propanamine, 1-propanamine, n,n-diethyl-1,1,2,3,3,3-hexafluoro, n,n-diethyl-1,1,2,3,3,3-hexafluoropropyl amine, ishikawa s reagent, acmc-20ak9c, ishikawa's reagant | |
CCN(CC)C(C(C(F)(F)F)F)(F)F | |
223.16 | |
223.16 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- N,N-Diethyl-1,1,2,3,3,3-hexafluoropropylamine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only