missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%

Product Code. 10183680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183680 100 g Glass bottle 100g
10772895 5 g Glass bottle 5g
10011780 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183680 Supplier Thermo Scientific Acros Supplier No. 155691000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 996-50-9 | C7H19NSi | 145.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 996-50-9
Molecular Formula C7H19NSi
Molecular Weight (g/mol) 145.32
MDL Number MFCD00009040
InChI Key JOOMLFKONHCLCJ-UHFFFAOYSA-N
Synonym n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
PubChem CID 70454
ChEBI CHEBI:85070
IUPAC Name N-ethyl-N-trimethylsilylethanamine
SMILES CCN(CC)[Si](C)(C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7600g/mL
Boiling Point 125°C to 126°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (C2H5)2NSi(CH3)3
Refractive Index 1.4103 to 1.4123
Quantity 100 g
Fieser 03,317; 04,544; 06,634
Specific Gravity 0.76
Solubility Information Solubility in water: decomposes
Formula Weight 145.32
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethyl-1,1,1-trimethylsilylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.