missing translation for 'onlineSavingsMsg'
Learn More

N,N-Didesmethyl Trimebutine-d5 Hydrochloride, TRC

Product Code. 30386405
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386405 1 mg
30381328 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30386405 Supplier Toronto Research Chemicals Supplier No. TRCD4413521MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1286624-16-5
Molecular Formula C20H20D5NO5 . HCl
Molecular Weight (g/mol) 400.9
Synonym 3,4,5-Trimethoxybenzoic Acid 2-Amino-2-phenylbutyl-d5 Ester Hydrochloride
SMILES Cl.[2H]C([2H])([2H])C([2H])([2H])C(N)(COC(=O)c1cc(OC)c(OC)c(OC)c1)c2ccccc2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H25NO5.ClH/c1-5-20(21,15-9-7-6-8-10-15)13-26-19(22)14-11-16(23-2)18(25-4)17(12-14)24-3;/h6-12H,5,13,21H2,1-4H3;1H/i1D3,5D2;
Chemical Name or Material N,N-Didesmethyl Trimebutine-d5 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.