missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Dicyclohexylurea, 98%, Thermo Scientific Chemicals
£42.60 - £138.00
Chemical Identifiers
CAS | 2387-23-7 |
---|---|
MDL Number | MFCD00003829 |
InChI Key | ADFXKUOMJKEIND-UHFFFAOYSA-N |
Synonym | n,n'-dicyclohexylurea, dicyclohexylurea, urea, n,n'-dicyclohexyl, unii-zv7823vvim, chembl1458, zv7823vvim, urea,3-dicyclohexyl, urea,n'-dicyclohexyl, n,n inverted exclamation marka-dicyclohexylurea, urea, 1,3-dicyclohexyl |
PubChem CID | 4277 |
IUPAC Name | 1,3-dicyclohexylurea |
SMILES | C1CCC(CC1)NC(=O)NC2CCCCC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10214190
|
Thermo Scientific Acros
113910250 |
25 g | Glass bottle |
£42.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10385790
|
Thermo Scientific
113911000 |
100 g | Glass bottle |
£138.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2387-23-7 | |
ADFXKUOMJKEIND-UHFFFAOYSA-N | |
4277 | |
C1CCC(CC1)NC(=O)NC2CCCCC2 |
MFCD00003829 | |
n,n'-dicyclohexylurea, dicyclohexylurea, urea, n,n'-dicyclohexyl, unii-zv7823vvim, chembl1458, zv7823vvim, urea,3-dicyclohexyl, urea,n'-dicyclohexyl, n,n inverted exclamation marka-dicyclohexylurea, urea, 1,3-dicyclohexyl | |
1,3-dicyclohexylurea |
Specifications
2387-23-7 | |
100.0 | |
Authentic | |
C13H24N2O | |
MFCD00003829 | |
n,n'-dicyclohexylurea, dicyclohexylurea, urea, n,n'-dicyclohexyl, unii-zv7823vvim, chembl1458, zv7823vvim, urea,3-dicyclohexyl, urea,n'-dicyclohexyl, n,n inverted exclamation marka-dicyclohexylurea, urea, 1,3-dicyclohexyl | |
C1CCC(CC1)NC(=O)NC2CCCCC2 | |
224.35 | |
224.35 | |
Liquid |
97.5 | |
231°C to 235°C | |
98% | |
C6H11NHCONHC6H11 | |
12, II, 11 | |
ADFXKUOMJKEIND-UHFFFAOYSA-N | |
1,3-dicyclohexylurea | |
4277 | |
98% | |
N, N'-Dicyclohexylurea, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 219-213-7
missing translation for 'tsca' : TSCA
RUO – Research Use Only