missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Dicyclohexylcarbodiimide, TRC

Product Code. 30385976
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30385976 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30385976

Brand: Toronto Research Chemicals TRCD43850010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Dicyclohexylcarbodiimide
CAS 538-75-0
Purity Grade Notes GC
Molecular Formula C13 H22 N2
Quantity 10 g
Synonym 1,3-Dicyclohexylcarbodiimide, Bis(cyclohexyl)carbodiimide, DCC, DCCD, DCCI, Dicyclohexylcarbodiimide, N,N’-Methanetetraylbis[cyclohexanamine], NSC 30022, NSC 53373, NSC 57182
SMILES C1CCC(CC1)N=C=NC2CCCCC2
Molecular Weight (g/mol) 206.33
Formula Weight 206.1783
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.