missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Dicyclohexylcarbodiimide Pentachlorophenol Complex, TRC

Product Code. 30386110
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386110 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386110 Supplier Toronto Research Chemicals Supplier No. TRCD439100100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Dicyclohexylcarbodiimide Pentachlorophenol Complex
CAS 15406-98-1
Molecular Formula C19 H23 Cl5 N2 O . 2 C6 H Cl5 O
Quantity 100 g
Synonym Carbamimidic acid, N,N'-dicyclohexyl-, 2,3,4,5,6-pentachlorophenyl ester, compd. with 2,3,4,5,6-pentachlorophenol (1:2), Carbamimidic acid, N,N'-dicyclohexyl-, pentachlorophenyl ester, compd. with pentachlorophenol (1:2) (9CI), Pseudourea, 1,3-dicyclohexyl-2-(pentachlorophenyl)-, compd. with pentachlorophenol (1:2) (8CI), Phenol, pentachloro-, compd. with 1,3-dicyclohexyl-2-(pentachlorophenyl)pseudourea (2:1) (8CI), Kovacs Complex, N,N'-Dicyclohexylcarbodiimide pentachlorophenol complex
InChI Formula InChI=1S/C19H23Cl5N2O.2C6HCl5O/c20-13-14(21)16(23)18(17(24)15(13)22)27-19(25-11-7-3-1-4-8-11)26-12-9-5-2-6-10-12;2*7-1-2(8)4(10)6(12)5(11)3(1)9/h11-12H,1-10H2,(H,25,26);2*12H
SMILES Oc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl.Oc2c(Cl)c(Cl)c(Cl)c(Cl)c2Cl.Clc3c(Cl)c(Cl)c(O\C(=N\C4CCCCC4)\NC5CCCCC5)c(Cl)c3Cl
IUPAC Name 2,3,4,5,6-pentachlorophenol;(2,3,4,5,6-pentachlorophenyl) N,N'-dicyclohexylcarbamimidate
Molecular Weight (g/mol) 1005.34
Formula Weight 997.7193
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.