missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylcarbodiimide, 99%

Product Code. 10498083 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
25 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
1kg
250g
25kg
5kg
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10498083

Brand: Thermo Scientific Acros 113900050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg | CAS: 538-75-0 | C13H22N2 | 206.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-75-0
Molecular Formula C13H22N2
Molecular Weight (g/mol) 206.33
MDL Number MFCD00011659
InChI Key QOSSAOTZNIDXMA-UHFFFAOYSA-N
Synonym dicyclohexylcarbodiimide, n,n'-dicyclohexylcarbodiimide, dccd, 1,3-dicyclohexylcarbodiimide, dcci, carbodicyclohexylimide, bis cyclohexyl carbodiimide, dcc, n,n-dicyclohexylcarbodiimide, carbodiimide, dicyclohexyl
PubChem CID 10868
ChEBI CHEBI:53090
IUPAC Name N,N'-dicyclohexylmethanediimine
SMILES C1CCC(CC1)N=C=NC2CCCCC2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 33.0°C to 35.0°C
Color White to Yellow
Boiling Point 122.0°C to 124.0°C (6.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Linear Formula C6H11N=C=NC6H11
Quantity 5 kg
Fieser 01,231; 02,126; 03,91; 04,141; 05,206; 06,174; 07,100; 08,162; 09,156; 10,142; 11,173; 13,333; 14,131; 15,131; 16,128
Merck Index 15,3107
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents,100g/l dichloromethane
Viscosity 5.81 mPa.s (55°C)
Formula Weight 206.33
Percent Purity 99%
Physical Form Crystalline Mass or Waxy Solid
Chemical Name or Material N,N′-Dicyclohexylcarbodiimide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.