missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicyclohexylcarbamimidothioic acid (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl Ester Dihydrochloride, TRC

Product Code. 30387611
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387611 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387611 Supplier Toronto Research Chemicals Supplier No. TRCD43845025MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Dicyclohexylcarbamimidothioic acid (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl Ester Dihydrochloride
CAS 1092776-63-0
Molecular Formula C21H34N4S2 . 2(HCl)
Quantity 25 mg
Synonym Carbamimidothioic acid, N,N'-dicyclohexyl-, (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl ester, hydrochloride (1:2) (ACI), IT1t, N,N'-Dicyclohexylcarbamimidothioic acid (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl ester dihydrochloride, (6,6-Dimethyl-5,6-dihydroimidazo[2,1-b]thiazol-3-yl)methyl N,N'-dicyclohexylcarbamimidothioate dihydrochloride
InChI Formula InChI=1S/C21H34N4S2.2ClH/c1-21(2)15-25-18(14-27-20(25)24-21)13-26-19(22-16-9-5-3-6-10-16)23-17-11-7-4-8-12-17;;/h14,16-17H,3-13,15H2,1-2H3,(H,22,23);2*1H
SMILES Cl.Cl.CC1(C)CN2C(=CSC2=N1)CS\C(=N\C3CCCCC3)\NC4CCCCC4
Molecular Weight (g/mol) 479.6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.