missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dicyclohexyl-2-benzothiazolsulfene Amide, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Dicyclohexyl-2-benzothiazolsulfene Amide |
| CAS | 4979-32-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H26 N2 S2 |
| Quantity | 1 g |
| Synonym | N,N-Dicyclohexyl-2-benzothiazolesulfenamide (ACI), 2-Benzothiazolyl-N,N-dicyclohexylsulfenamide, Accel DZ-G, Accelerator DZ, DCBS, DTs, M 181, Meramid DCH, Mercure DCBS, N,N-Dicyclohexyl-1,3-benzothiazole-2-sulfenamide, N,N-Dicyclohexyl-2-benzothiazolylsulfenamide, N,N-Dicyclohexyl-2-mercaptobenzothiazole sulfenamide, Nocceler DZ, Nocceler DZ-G, Rhodifax 30, SA-DTs, Sanceler DZ, Sanceler DZ-G, Santocure DCBS, Soxinol DZ, Sulfenamide DTs, Vulkacit DZ |
| InChI Formula | InChI=1S/C19H26N2S2/c1-3-9-15(10-4-1)21(16-11-5-2-6-12-16)23-19-20-17-13-7-8-14-18(17)22-19/h7-8,13-16H,1-6,9-12H2 |
| SMILES | C1CCC(CC1)N(Sc2nc3ccccc3s2)C4CCCCC4 |
| IUPAC Name | N-(1,3-benzothiazol-2-ylsulfanyl)-N-cyclohexyl-cyclohexanamine |
| Molecular Weight (g/mol) | 346.55 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?