missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dicarbamoylhydrazine, 98%, Thermo Scientific™

Product Code. 11385159
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385159 10 g 10g
11395159 50 g 50g
11305169 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385159 Supplier Thermo Scientific Alfa Aesar Supplier No. A18260.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-21-4
Molecular Formula C2H6N4O2
Molecular Weight (g/mol) 118.096
MDL Number MFCD00025398
InChI Key ULUZGMIUTMRARO-UHFFFAOYSA-N
Synonym biurea, 1,2-hydrazinedicarboxamide, ureidourea, bicarbamamide, bicarbamimidic acid, hydrazodicarbonamide, hydrazodicarboxamide, hydrazine-1,2-dicarboxamide, hydrazocarbonamide, urea, ureido
PubChem CID 8039
IUPAC Name (carbamoylamino)urea
SMILES C(=O)(N)NNC(=O)N

Specifications

Melting Point 239°C to 241°C (decomposition)
Quantity 10 g
Formula Weight 118.1
Percent Purity 98%
Chemical Name or Material N,N'-Dicarbamoylhydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.