missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibutylaniline, 97%

Product Code. 11383017
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11383017

Brand: Thermo Scientific Alfa Aesar L13390.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 613-29-6 | C14H23N | 205.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-29-6
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.35
MDL Number MFCD00035799
InChI Key FZPXKEPZZOEPGX-UHFFFAOYSA-N
Synonym benzenamine, n,n-dibutyl, n,n-di-n-butylaniline, dibutylaniline, aniline, n,n-dibutyl, dibutyl-phenyl-amine, dibutylphenylamine, n,n-dibutyl aniline, n-phenyldibutylamine, n,n-dibutylbenzenamine, dibutylamine, n-phenyl
PubChem CID 61154
IUPAC Name N,N-dibutylaniline
SMILES CCCCN(CCCC)C1=CC=CC=C1

Specifications

Density 0.906
Boiling Point 269°C to 270°C
Flash Point >110°C (230°F)
Refractive Index 1.518
Quantity 5 g
UN Number UN2810
Beilstein 2210422
Formula Weight 205.35
Percent Purity 97%
Chemical Name or Material N,N-Dibutylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.