missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diboc 1-O-t-Butyl 6-Dehydroxy L-Glutamic Acid, TRC

Product Code. 30386228
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386228 10 mg 10mg
30378206 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386228 Supplier Toronto Research Chemicals Supplier No. TRCD41950510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Diboc 1-O-t-Butyl 6-Dehydroxy L-Glutamic Acid
CAS 1480890-33-2
Molecular Formula C20H35NO7
Quantity 10 mg
Synonym N,N-Bis[(1,1-dimethylethoxy)carbonyl]-6-oxo-L-norleucine 1,1-Dimethylethyl Ester
InChI Formula InChI=1S/C20H35NO7/c1-18(2,3)26-15(23)14(12-10-11-13-22)21(16(24)27-19(4,5)6)17(25)28-20(7,8)9/h13-14H,10-12H2,1-9H3/t14-/m0/s1
SMILES CC(C)(C)OC(=O)[C@H](CCCC=O)N(C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
IUPAC Name tert-butyl (2S)-2-[bis[(2-methylpropan-2-yl)oxycarbonyl]amino]-6-oxohexanoate
Molecular Weight (g/mol) 401.49
Formula Weight 401.2414
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.