missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzylethylenediamine Diacetate Salt, TRC

Product Code. 30137275
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137275 100 g 100g
30138717 1 g 1g
17944285 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137275 Supplier Toronto Research Chemicals Supplier No. TRCD417190100G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-75-8
Molecular Formula C16 H20 N2 . 2 C2 H4 O2
Molecular Weight (g/mol) 360.45
Synonym N,N'-Dibenzylethylenediamine diacetate, 1,2-Ethanediamine, N1,N2-bis(phenylmethyl)-, acetate (1:2), 1,2-Ethanediamine, N,N'-bis(phenylmethyl)-, diacetate (9CI), Ethylenediamine, N,N'-dibenzyl-, diacetate (6CI,7CI,8CI), Benzathine diacetate, NSC 33274, NSC 81591
IUPAC Name acetic acid;N,N'-dibenzylethane-1,2-diamine
SMILES CC(=O)O.CC(=O)O.C(CNCc1ccccc1)NCc2ccccc2

Specifications

Quantity 100 g
InChI Formula InChI=1S/C16H20N2.2C2H4O2/c1-3-7-15(8-4-1)13-17-11-12-18-14-16-9-5-2-6-10-16;2*1-2(3)4/h1-10,17-18H,11-14H2;2*1H3,(H,3,4)
Formula Weight 360.2049
Chemical Name or Material N,N'-Dibenzylethylenediamine Diacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.