missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dibenzylaniline, 99%

Product Code. 15484767
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484767 25 g 25g
15494767 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484767 Supplier Thermo Scientific Alfa Aesar Supplier No. H32091.14
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 91-73-6 | C20H19N | 273.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-73-6
Molecular Formula C20H19N
Molecular Weight (g/mol) 273.38
MDL Number MFCD00022015
InChI Key ISGXOWLMGOPVPB-UHFFFAOYSA-N
Synonym dibenzylaniline, n-phenyldibenzylamine, aniline, n,n-dibenzyl, benzenemethanamine, n-phenyl-n-phenylmethyl, dibenzylamine, n-phenyl, benzenamine, n,n-bis phenylmethyl, dibenzylaniline, n,n-bis phenylmethyl, dibenzylanilin, bisbenzylphenylamine, aniline, dibenzyl
PubChem CID 66681
IUPAC Name N,N-dibenzylaniline
SMILES C(N(CC1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 69°C to 71°C
Boiling Point 180°C (1 mmHg)
Quantity 25 g
Formula Weight 273.37
Percent Purity 99%
Chemical Name or Material N,N-Dibenzylaniline
compliance-icons
Product Identifier
  • N,N-Dibenzylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: N-phenyl-N-(phenylmethyl)-benzenemethanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.