missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylhydrazine, 98%

Product Code. p-7021166
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475393 10 g 10g
11485393 50 g 50g
11495393 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475393 Supplier Thermo Scientific Alfa Aesar Supplier No. A13393.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3148-73-0 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 3148-73-0
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00008673
InChI Key ZLHNYIHIHQEHJQ-UHFFFAOYSA-N
Synonym n,n'-diacetylhydrazine, 1,2-diacetylhydrazine, acetic acid, 2-acetylhydrazide, diacetylahydrazine, diacetylhydrazine, sym-diacetylhydrazine, unii-ymr3zf0q4m, hydrazine, 1,2-diacetyl, 1.2-diacetylhydrazine, ymr3zf0q4m
PubChem CID 72884
ChEBI CHEBI:80616
IUPAC Name N'-acetylacetohydrazide
SMILES CC(=O)NNC(=O)C

Especificaciones

Melting Point 138°C to 142°C
Boiling Point 209°C (15 mmHg)
Quantity 10 g
Beilstein 1754018
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material N,N'-Diacetylhydrazine

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.