missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Diacetyl-L-cystine Bis(tert-Butyl) Diester, TRC

Product Code. 30350273
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350273 10 mg 10mg
30358271 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350273 Supplier Toronto Research Chemicals Supplier No. TRCD31251510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1079950-07-4
Molecular Formula C18 H32 N2 O6 S2
Molecular Weight (g/mol) 436.59
Synonym N,N'-Diacetyl-L-cystine 1,1’-Bis(1,1-dimethylethyl) Ester
IUPAC Name tert-butyl (2S)-2-acetamido-3-[[(2S)-2-acetamido-3-[(2-methylpropan-2-yl)oxy]-3-oxopropyl]disulfanyl]propanoate
SMILES CC(=O)N[C@H](CSSC[C@@H](NC(=O)C)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C

Specifications

InChI Formula InChI=1S/C18H32N2O6S2/c1-11(21)19-13(15(23)25-17(3,4)5)9-27-28-10-14(20-12(2)22)16(24)26-18(6,7)8/h13-14H,9-10H2,1-8H3,(H,19,21)(H,20,22)/t13-,14-/m1/s1
Quantity 10 mg
Formula Weight 436.1702
Shipping Condition Room Temperature
Chemical Name or Material N,N'-Diacetyl-L-cystine Bis(tert-Butyl) Diester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.