missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-tert-butylethylenediamine, 98%

Product Code. 11311787
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311787 25 g 25g
11301787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11311787 Supplier Thermo Scientific Alfa Aesar Supplier No. L04972.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4062-60-6 | C10H24N2 | 172.316 g/mol

N,N'-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-tert-butylethylenediamine is used as a primary and secondary intermediate. It is also used as a pharmaceutical intermediate.

Solubility
Soluble (33 g/L) (25°C),

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4062-60-6
Molecular Formula C10H24N2
Molecular Weight (g/mol) 172.316
MDL Number MFCD00014996
InChI Key KGHYGBGIWLNFAV-UHFFFAOYSA-N
Synonym n,n'-di-tert-butylethylenediamine, 1,2-bis tert-butylamino ethane, n,n'-di-t-butylethylenediamine, n1,n2-di-tert-butylethane-1,2-diamine, unii-6wai8u5v0w, n,n'-bis tert-butyl ethylenediamine, 1,2-ethanediamine, n,n'-bis 1,1-dimethylethyl, n,n'-di-tert-butyl-ethylenediamine, 6wai8u5v0w, n,n'-bis 1,1-dimethylethyl-1,2-ethanediamine
PubChem CID 77680
IUPAC Name N,N'-ditert-butylethane-1,2-diamine
SMILES CC(C)(C)NCCNC(C)(C)C

Specifications

Density 0.801
Boiling Point 132°C (100 mmHg)
Flash Point 62°C (143°F)
Linear Formula (CH3)3CNHCH2CH2NHC(CH3)3
Refractive Index 1.43
Quantity 25 g
UN Number UN2735
Beilstein 1738432
Solubility Information Soluble (33g/L) (25°C),
Formula Weight 172.32
Percent Purity 98%
Chemical Name or Material N,N'-Di-tert-butylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.