missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Di-n-butylurea, 98%

Product Code. 11302406
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302406 5 g 5g
11312406 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11302406 Supplier Thermo Scientific Alfa Aesar Supplier No. L06165.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1792-17-2 | C9H20N2O | 172.27 g/mol

N,N'-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Di-n-butylurea is used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1792-17-2
Molecular Formula C9H20N2O
Molecular Weight (g/mol) 172.27
MDL Number MFCD00015229
InChI Key AQSQFWLMFCKKMG-UHFFFAOYSA-N
Synonym n,n'-dibutylurea, n,n'-di-n-butylurea, urea, n,n'-dibutyl, 1,3-dibutyl urea, unii-j2hug1i10w, j2hug1i10w, n-butyl butylamino carboxamide, nn'-dibutylurea, urea,n'-dibutyl, 1,3-dibutyl-urea
PubChem CID 74523
IUPAC Name 1,3-dibutylurea
SMILES CCCCNC(=O)NCCCC

Specifications

Melting Point 69°C to 75°C
Boiling Point 115°C (0.03 mmHg)
Quantity 5 g
Beilstein 1704165
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material N,N'-Di-n-butylurea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.