missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-butyl-2,6-difluorobenzamide, 97%, Thermo Scientific™

Catalog Number 15435968
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
959048-95-4
C15H21F2NO
269.34
MFCD22988987
BTXNGJCNHCDPDU-UHFFFAOYSA-N
benzamide, n,n-dibutyl-2,6-difluoro, n,n-di-n-butyl-2,6-difluorobenzamide
533350
N,N-dibutyl-2,6-difluorobenzamide
CCCCN(CCCC)C(=O)C1=C(F)C=CC=C1F
This item is not returnable. View return policy
959048-95-4
C15H21F2NO
269.34
MFCD22988987
BTXNGJCNHCDPDU-UHFFFAOYSA-N
benzamide, n,n-dibutyl-2,6-difluoro, n,n-di-n-butyl-2,6-difluorobenzamide
533350
N,N-dibutyl-2,6-difluorobenzamide
CCCCN(CCCC)C(=O)C1=C(F)C=CC=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 269.33
Percent Purity 97%
Chemical Name or Material N,N-Di-n-butyl-2,6-difluorobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'