missing translation for 'onlineSavingsMsg'
Learn More

N-N-Butyldiethanolamine, 98%

Product Code. 10237023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10237023 2.5 L Glass bottle 2.5 L
10031740 250 mL Glass bottle 250mL
10276100 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10237023 Supplier Thermo Scientific Acros Supplier No. 154360025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-79-4 | C8H19NO2 | 161.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-79-4
Molecular Formula C8H19NO2
Molecular Weight (g/mol) 161.24
MDL Number MFCD00002856
InChI Key GVNHOISKXMSMPX-UHFFFAOYSA-N
Synonym n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
PubChem CID 7620
IUPAC Name 2-[butyl(2-hydroxyethyl)amino]ethanol
SMILES CCCCN(CCO)CCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Brown to Brown-Red
Density 0.9680g/mL
Boiling Point 264.0°C to 278.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3N(CH2CH2OH)2
Refractive Index 1.4615 to 1.4635
Quantity 2.5 L
Beilstein 04, 285
Specific Gravity 0.968
Solubility Information Solubility in water: miscible
Formula Weight 161.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-N-Butyldiethanolamine
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.