missing translation for 'onlineSavingsMsg'
Learn More

N-N-Butyldiethanolamine, 98%, Thermo Scientific Chemicals

Catalog Number 10031740
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
102-79-4
C8H19NO2
161.24
MFCD00002856
GVNHOISKXMSMPX-UHFFFAOYSA-N
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
7620
2-[butyl(2-hydroxyethyl)amino]ethanol
CCCCN(CCO)CCO
This item is not returnable. View return policy
102-79-4
C8H19NO2
161.24
MFCD00002856
GVNHOISKXMSMPX-UHFFFAOYSA-N
n-butyldiethanolamine, butyldiethanolamine, ethanol, 2,2'-butylimino bis, bide, 2,2'-butylimino diethanol, n,n-bis 2-hydroxyethyl butylamine, butylbis 2-hydroxyethyl amine, n-butyl-2,2'-iminodiethanol, 2,2-butylimino diethanol, n-butyl-n,n-bis hydroxyethyl amine
7620
2-[butyl(2-hydroxyethyl)amino]ethanol
CCCCN(CCO)CCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -70.0°C
Color Brown to Brown-Red
Density 0.9680g/mL
Boiling Point 264.0°C to 278.0°C
Flash Point 143°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3N(CH2CH2OH)2
Refractive Index 1.4615 to 1.4635
Quantity 250 mL
Beilstein 04, 285
Specific Gravity 0.968
Solubility Information Solubility in water: miscible
Formula Weight 161.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-N-Butyldiethanolamine
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'