missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)thiophosphoric Triamide, TRC

Product Code. 17953295
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953295 10 g 10g
17933645 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17953295 Supplier Toronto Research Chemicals Supplier No. TRCB69400010G
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94317-64-3
Molecular Formula C4H14N3PS
Molecular Weight (g/mol) 167.21
Synonym Phosphorothioic triamide, N-butyl-, Phosphorothioic triamide, butyl- (6CI,9CI), N-Butylphosphorothioic triamide, Agrotain, Agrotain DC, Agrotain Plus, ArmU, N-(n-Butyl)phosphorothioic triamide, N-(n-Butyl)thiophosphoric triamide, N-Butylthiophosphoric triamide
IUPAC Name N-diaminophosphinothioylbutan-1-amine
SMILES CCCCNP(=S)(N)N

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H14N3PS/c1-2-3-4-7-8(5,6)9/h2-4H2,1H3,(H5,5,6,7,9)
Formula Weight 167.0646
Chemical Name or Material N-(n-Butyl)thiophosphoric Triamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.