missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)phthalimide, 99%

Product Code. 11448646
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448646 5 g 5g
11458646 25 g 25g
11468646 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448646 Supplier Thermo Scientific Alfa Aesar Supplier No. A16020.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1515-72-6 | C12H13NO2 | 203.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1515-72-6
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
MDL Number MFCD00039695
InChI Key DLKDEVCJRCPTLN-UHFFFAOYSA-N
Synonym n-butylphthalimide, 2-butylisoindoline-1,3-dione, n-n-butyl phthalimide, 1-phthalimidobutane, phthalimide, n-butyl, 1h-isoindole-1,3 2h-dione, 2-butyl, unii-5th1dkt35e, 2-butyl-1h-isoindole-1,3 2h-dione, 5th1dkt35e, 2-butylbenzo c azoline-1,3-dione
PubChem CID 73812
IUPAC Name 2-butylisoindole-1,3-dione
SMILES CCCCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 32°C to 35°C
Boiling Point 120°C to 123°C (5 mmHg)
Quantity 5 g
Beilstein 147784
Formula Weight 203.24
Percent Purity 99%
Chemical Name or Material N-(n-Butyl)phthalimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.