missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)phosphoric Triamide, TRC

Product Code. 30132641
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132641 1 g 1g
30132402 250 mg 250mg
17914715 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132641 Supplier Toronto Research Chemicals Supplier No. TRCB6936501G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25316-39-6
Molecular Formula C4 H14 N3 O P
Molecular Weight (g/mol) 151.15
Synonym Phosphoric triamide, butyl- (6CI,8CI,9CI), N-Butylphosphoric triamide, N-Butylphosphoric Triamide
IUPAC Name N-diaminophosphorylbutan-1-amine
SMILES CCCCNP(=O)(N)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H14N3OP/c1-2-3-4-7-9(5,6)8/h2-4H2,1H3,(H5,5,6,7,8)
Formula Weight 151.0874
Chemical Name or Material N-(n-Butyl)phosphoric Triamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.