missing translation for 'onlineSavingsMsg'
Learn More

N-(n-Butyl)ethylenediamine, 97%

Product Code. 11313067
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313067 25 g 25g
11303067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313067 Supplier Thermo Scientific Alfa Aesar Supplier No. L13437.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 19522-69-1 | C6H16N2 | 116.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19522-69-1
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00025602
InChI Key DFPGBRPWDZFIPP-UHFFFAOYSA-N
Synonym n-butylethylenediamine, 2-butylamino ethylamine, n1-butylethane-1,2-diamine, 2-aminoethyl butyl amine, 1,2-ethanediamine,n1-butyl, 2-butylamino-ethylamine, 2-n-butylaminoethylamine, acmc-1bs5m, n-butyl-1,2-ethanediamine, n-butylethane-1,2-diamine
PubChem CID 519668
IUPAC Name N'-butylethane-1,2-diamine
SMILES CCCCNCCN

Specifications

Density 0.836
Boiling Point 170°C to 171°C
Flash Point 59°C (138°F)
Refractive Index 1.443
Quantity 25 g
UN Number UN2734
Beilstein 1732700
Sensitivity Air Sensitive
Formula Weight 116.21
Percent Purity 97%
Chemical Name or Material N-(n-Butyl)ethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.