missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(trimethylsilyl)urea, 98+%

Product Code. 11344544
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344544 50 g 50g
11354544 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344544 Supplier Thermo Scientific Alfa Aesar Supplier No. L01301.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18297-63-7 | C7H20N2OSi2 | 204.42 g/mol

N,N'-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Bis(trimethylsilyl)urea is used as a reagent for the silylation of alcohols and carboxylic acids.

Solubility
Reacts with water.

Notes
Moisture Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18297-63-7
Molecular Formula C7H20N2OSi2
Molecular Weight (g/mol) 204.42
MDL Number MFCD00008260
InChI Key MASDFXZJIDNRTR-UHFFFAOYSA-N
Synonym 1,3-bis trimethylsilyl urea, n,n'-bis trimethylsilyl urea, bis trimethylsilyl urea, urea, n,n'-bis trimethylsilyl, n,n'-bis-trimethylsilyl-urea, urea, 1,3-bis trimethylsilyl, bsu, urea, bis-tms, hexamethyl disilaurea
PubChem CID 87562
IUPAC Name 1,3-bis(trimethylsilyl)urea
SMILES C[Si](C)(C)NC(=O)N[Si](C)(C)C

Specifications

Melting Point ∽230°C (decomposition)
Quantity 50 g
Beilstein 1937452
Solubility Information Reacts with water.
Formula Weight 204.42
Percent Purity ≥98%
Sensitivity Moisture sensitive
Chemical Name or Material N,N'-Bis(trimethylsilyl)urea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.