missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Bis(salicylidene)ethylenediamine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis(salicylidene)ethylenediamine |
| CAS | 94-93-9 |
| Molecular Formula | C16H16N2O2 |
| Quantity | 5 g |
| Synonym | 2,2'-[1,2-Ethanediylbis(nitrilomethylidyne)]bisphenol, 1,2-Bis(salicylideneamino)ethane, 1,2-Bis[[(2-hydroxyphenyl)methylene]amino]ethane, 1,2-Ethanedi(salicylaldimine), N,N'-bis[(2-hydroxyphenyl)methylene]-1,2-ethanediamine, 1,6-Bis(2-hydroxyphenyl)-2,5-diazahexa-1,5-diene, Bis(salicylaldehyde)ethylenediimine, Di(salicylidene)ethylenediamine, Disalicylidene-1,2-ethanediamine, N,N'-Bis(2-hydroxybenzylidene)-1,2-ethanediamine, N,N'-Bis(2-hydroxybenzylidene)ethylenediamine, N,N'-Bis(o-hydroxybenzylidene)ethylenediamine, N,N'-Bis(salicylidene)-1,2-ethanediamine, N,N'-Bis(salicylidene)ethylene-1,2-diamine, N,N'-Di(salicylidene)ethylenediamine, N,N'-Disalicylidene-1,2-diaminoethane, N,N'-Disalicylidene-1,2-ethanediamine, N,N'-Ethylenebis(salicylaldimine), N,N'-Ethylenebis(salicylideneimine), NSC 2079, Salen, Salicylaldehyde Ethylenediamine Bisimine, Salicylaldehyde Ethylenediamine Diimine, α,α'-(Ethylenedinitrilo)di-o-cresol |
| InChI Formula | InChI=1S/C16H16N2O2/c19-15-7-3-1-5-13(15)11-17-9-10-18-12-14-6-2-4-8-16(14)20/h1-8,11-12,19-20H,9-10H2/b17-11+,18-12+ |
| SMILES | OC1=C(/C=N/CC/N=C/C2=C(O)C=CC=C2)C=CC=C1 |
| IUPAC Name | 2,2'-((1E,1'E)-(ethane-1,2-diylbis(azaneylylidene))bis(methaneylylidene))diphenol |
| Molecular Weight (g/mol) | 268.31 |
| Formula Weight | 268.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction