missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Bis(salicylidene)ethylenediamine, 98%

Product Code. 11421588
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421588 500 g 500g
11401588 25 g 25g
11411588 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421588 Supplier Thermo Scientific Alfa Aesar Supplier No. A13885.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-93-9 | C16H16N2O2 | 268.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-93-9
Molecular Formula C16H16N2O2
Molecular Weight (g/mol) 268.32
MDL Number MFCD00002244
InChI Key RQHVNNWVDLRULK-XSYHWHKQSA-N
Synonym salen, salen unspecified, disalicylaldehyde ethylenediamine, disalicylidene-1,2-ethanediamine, n,n'-disalicylidene ethylenediamine, ethylenediamine, n,n'-disalicylidene, n,n'-bis salicylidene ethylenediamine, o-cresol, alpha,alpha-ethylenediiminodi, alpha,alpha'-ethylenedinitrilodi-o-cresol, disalicylalethylenediamine
PubChem CID 5464119
IUPAC Name (6Z)-6-[[2-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]ethylamino]methylidene]cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C\NCCN\C=C1\C=CC=CC1=O

Specifications

Melting Point 129°C to 131°C
Quantity 500 g
Beilstein 535296
Merck Index 14,8322
Formula Weight 268.32
Percent Purity 98%
Chemical Name or Material N,N'-Bis(salicylidene)ethylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.